missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,8,8-Heptamethylnonane, 98%

Product Code. 10589043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10589043 250 mL Glass bottle 250mL
10275960 50 mL Glass bottle 50mL
11904401 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10589043 Supplier Thermo Scientific Acros Supplier No. 156252500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 4390-04-9 | C16H34 | 226.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4390-04-9
Molecular Formula C16H34
Molecular Weight (g/mol) 226.44
MDL Number MFCD00008856
InChI Key VCLJODPNBNEBKW-UHFFFAOYNA-N
Synonym isocetane, nonane, 2,2,4,4,6,8,8-heptamethyl, cyprane, pubchem16042, acmc-2097fb, dsstox_cid_30668, dsstox_gsid_52101, ksc492g9j, 2,4,4,6,8,8-heptamethylnonane, nonane,2,4,4,6,8,8-heptamethyl
PubChem CID 20414
IUPAC Name 2,2,4,4,6,8,8-heptamethylnonane
SMILES CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.7900g/mL
Boiling Point 240.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH2CH(CH3)CH2C(CH3)2CH2C(CH3)3
Refractive Index 1.4380 to 1.4400
Quantity 250 mL
Specific Gravity 0.79
Solubility Information Solubility in water: insoluble
Formula Weight 226.44
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,8,8-Heptamethylnonane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,4,4,6,8,8-Heptamethylnonane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H332-Harmful if inhaled.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.