missing translation for 'onlineSavingsMsg'
Learn More

2-(2-(4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl)ethoxy)ethanol, TRC

Product Code. 17991705
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991705 25 mg 25mg
17921705 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991705 Supplier Toronto Research Chemicals Supplier No. TRCD41710525MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 848814-27-7
Molecular Formula C21 H27 N3 O3 S
Molecular Weight (g/mol) 401.52
Synonym 1-[2-[(2-Aminophenyl)thio]benzoyl]-4-[2-(2-hydroxyethoxy)ethyl]-piperazine, [2-[(2-Aminophenyl)thio]phenyl][4-[2-(2-hydroxyethoxy)ethyl]-1-piperazinyl]-methanone, Quetiapine EP Impurity F
IUPAC Name [2-(2-aminophenyl)sulfanylphenyl]-[4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]methanone
SMILES Nc1ccccc1Sc2ccccc2C(=O)N3CCN(CCOCCO)CC3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H27N3O3S/c22-18-6-2-4-8-20(18)28-19-7-3-1-5-17(19)21(26)24-11-9-23(10-12-24)13-15-27-16-14-25/h1-8,25H,9-16,22H2
Formula Weight 401.1773
Chemical Name or Material 2-(2-(4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl)ethoxy)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.