missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate

Product Code. 11303938
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11303938 500 g 500g
11393928 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303938 Supplier Thermo Scientific Alfa Aesar Supplier No. 040366.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25265-77-4 | C12H24O3 | 216.321 g/mol

Employed In pharma industry. As Coalescent for latex paints. Finds applications in Coatings, Nail care, Printing inks, Solvents for cosmetics and personal care, plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25265-77-4
Molecular Formula C12H24O3
Molecular Weight (g/mol) 216.321
MDL Number MFCD00148967
InChI Key DAFHKNAQFPVRKR-UHFFFAOYSA-N
Synonym texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
PubChem CID 6490
IUPAC Name (3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)O

Specifications

Melting Point -50°C
Color Colorless
Density 0.950 g/mL
Boiling Point 254°C to 261°C
Refractive Index 1.442
Quantity 500 g
Solubility Information Soluble in water 900mg/L.
Formula Weight 216.32
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.