missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate (TXIB) |
| CAS | 6846-50-0 |
| Molecular Formula | C16 H30 O4 |
| Quantity | 1 g |
| Synonym | Propanoic acid','2-methyl-','1,1'-[2,2-dimethyl-1-(1-methylethyl)-1,3-propanediyl] ester (ACI), 1,3-Pentanediol','2,2,4-trimethyl-','diisobutyrate (8CI), Isobutyric acid','1-isopropyl-2,2-dimethyltrimethylene ester (7CI','8CI), Propanoic acid','2-methyl-','2,2-dimethyl-1-(1-methylethyl)-1,3-propanediyl ester (9CI), 1-Isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-Trimethyl-1,3-pentanediol 2-methylpropanoic acid diester, 2,2,4-Trimethyl-1,3-pentanediol bis(isobutyrate), 2,2,4-Trimethyl-1,3-pentanediol di(isopropylformate), 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 2,2,4-Trimethyl-1,3-pentanediyl diisobutyrate, 2,2,4-Trimethylpentane 1,3-diisobutanoate, 2,2-Dimethyl-1-(1-methylethyl)-1,3-propanediyl bis(2-methylpropanoate), [2,2,4-Trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate, C 16, CS 16, Eastman TXIB, Kodaflex TXIB, Kyowanol D, Kyowanol M, NX 800, OE 300, Optifilm Enhancer 300, RTC 16, Texanol isobutyrate, Texanol OE 300, TXIB |
| InChI Formula | InChI=1S/C16H30O4/c1-10(2)13(20-15(18)12(5)6)16(7,8)9-19-14(17)11(3)4/h10-13H,9H2,1-8H3 |
| SMILES | CC(C)C(OC(=O)C(C)C)C(C)(C)COC(=O)C(C)C |
| IUPAC Name | [2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate |
| Molecular Weight (g/mol) | 286.41 |
| Formula Weight | 286.2144 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?