missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate, 99%, Honeywell™

Product Code. 15635810 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Glass bottle
Stainless steel can
Unit Size:
1 L
20 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15635810 20 L Stainless steel can 20 L
15625810 1 L Glass bottle 1 L
15618430 4 L Glass bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15635810 Supplier Honeywell Research Chemicals Supplier No. 53822120L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mixture of isomers, 99%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25265-77-4
Molecular Formula C12H24O3
Molecular Weight (g/mol) 216.321
InChI Key DAFHKNAQFPVRKR-UHFFFAOYSA-N
Synonym texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
PubChem CID 6490
IUPAC Name (3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)O

Specifications

Melting Point -50°C
Color Colorless
Density 0.95g/mL at 25°C
Boiling Point 255°C
Ignition Point 388°C
Packaging Stainless steel can
Linear Formula (CH3)2CHCH[O(R)]C(CH3)2CH2O(R), R=-COCH(CH3)2 or H
Quantity 20 L
UN Number NONH for all modes of transport
Formula Weight 216.32g/mol
Vapor Pressure 0.01mmHg (20°C)
Percent Purity 99%
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.