missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol, TRC

Product Code. 30456939
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456939 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456939 Supplier Toronto Research Chemicals Supplier No. TRCT796570100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol
CAS 144-19-4
Molecular Formula C8 H18 O2
Quantity 100 g
Synonym 2,2,4-Trimethyl-1,3-pentanediol, 2,2,4-Trimethylpentan-1,3-diol, Nexcoat 795, NSC 6368, TMPD (alcohol), 2,2,4-Trimethyl-1,3-pentanediol, NSC 6368, Nexcoat 795, TMPD (Alcohol)
InChI Formula InChI=1S/C8H18O2/c1-6(2)7(10)8(3,4)5-9/h6-7,9-10H,5H2,1-4H3
SMILES CC(C)C(O)C(C)(C)CO
IUPAC Name 2,2,4-trimethylpentane-1,3-diol
Molecular Weight (g/mol) 146.227
Formula Weight 146.1307
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.