missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, 99.5%, for HPLC

Product Code. p-216651 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265371 1 L Glass Bottle 1 L
10762291 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10265371 Supplier Thermo Scientific Supplier No. 268810010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

For HPLC | CAS: 540-84-1 | C8H18 | 114.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107.0°C
Absorbance (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 240nm, 0.005 max. at 250nm, 0.02 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.4 max. at 200nm
CAS Min % 99.4
Color Colorless to Yellow
Acidity 0.0002 meq/g max.
CAS Max % 100.0
Density 0.6900g/mL
Boiling Point 98.0°C to 99.0°C
Quantity 1 L
Flash Point −12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3915
Assay Percent Range 99.5%
Residue after Evaporation 0.0002 % max.
Specific Gravity 0.69
Linear Formula (CH3)2CHCH2C(CH3)3
Viscosity 0.51 mPa.s (22°C)
Beilstein 01, IV, 439
Water 0.01% Max.
Merck Index 15, 5239
Green Features ACT Ecolabel (Gold)
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol, soluble in benzene, toluene, xylene, chloroform,, dmf, ether, ccl4 and cs2, miscible with acetone, soluble in oils except castor oil
Formula Weight 114.23
Percent Purity 99.5%
Grade HPLC
Chemical Name or Material 2, 2, 4-Trimethylpentane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2,4-Trimethylpentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.