missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, 99+%, pure, ACROS Organics™

Product Code. 10760682 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
2kg; Metal drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10760682 2kg; Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10760682 Supplier Thermo Scientific Acros Supplier No. 421970020

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

CAS 540-84-1
CAS Min % 99.0
Color Colorless
CAS Max % 100.0
Density 0.6900g/mL
Quantity 2 kg
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.3900 to 1.3920 (20°C, 589nm)
Assay Percent Range 99+%
Molecular Formula C8H18
Linear Formula (CH3)2CHCH2C(CH3)3
MDL Number MFCD00008943
Beilstein 01, IV, 439
Merck Index 15, 5239
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.232
PubChem CID 10907
ChEBI CHEBI:62805
Formula Weight 114.23
Percent Purity ≥99.0%
Grade Pure
Chemical Name or Material 2, 2, 4-Trimethylpentane, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.