missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ≥99%, Reagent Grade, Honeywell™

Product Code. p-7120045 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
20 L
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15602800

Brand: Honeywell Research Chemicals 258776500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Color Colorless
Density 0.692g/mL at 25°C
Boiling Point 98°C to 99°C
Quantity 500 mL
Explosion Limit 0.06
Flash Point −12°C (10.4°F)
Ignition Point 745°F
Packaging Glass Bottle
Physical Form Liquid
Linear Formula (CH3)2CHCH2C(CH3)3
Vapor Pressure 41mmHg (21°C)
UN Number UN1262
Beilstein 1696876
Formula Weight 114.23g/mol
Percent Purity ≥99%
Grade ReagentPlus
Chemical Name or Material 2,2,4-Trimethylpentane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.