missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, 99%

Product Code. 11438637
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438637 2500 mL 2500mL
11428637 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11438637 Supplier Thermo Scientific Alfa Aesar Supplier No. A19756.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 540-84-1 | C8H18 | 114.232 g/mol

2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.

Solubility
Miscible with benzene, toluene, xylene, chloroform, ether, carbon disulfide, carbon tetrachloride, DMF, acetone, heptane, and oils. Immiscible with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Density 0.692
Boiling Point 98°C to 99°C
Flash Point −7°C (19°F)
Odor Nearly Odorless
Linear Formula (CH3)2CHCH2C(CH3)3
Refractive Index 1.391
Quantity 2500 mL
UN Number UN1262
Beilstein 1696876
Merck Index 14,5193
Solubility Information Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,DMF,acetone,heptane,and oils. Immiscible with water.
Formula Weight 114.23
Percent Purity 99%
Chemical Name or Material 2,2,4-Trimethylpentane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2,4-Trimethylpentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: 2,2,4-Trimethylpentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.