missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ACS, 99+%

Product Code. 11400530
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400530 500 mL 500mL
11328287 1 L 1 L
11338287 4 x 1 L 4 L
11348287 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11400530 Supplier Thermo Scientific Alfa Aesar Supplier No. 031787.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 540-84-1 | C8H18 | 114.232 g/mol

Solvent, thinner, in determination of fuel octane numbers2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as a thinner. It is involved as a processing aid in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Color Colorless
Density 0.692
Boiling Point 98°C to 99°C
Flash Point -7°C (19°F)
Assay Percent Range >99%
Odor Nearly Odorless
Refractive Index 1.391
Quantity 500 mL
UN Number 1262
Beilstein 1696876
Solubility Information Soluble in benzene,toluene,xylene,chloroform,ether,CS2,CCl4,and DMF. Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane and oils. Immiscible with water.
Formula Weight 114.23
Vapor Pressure 15 hPa (11mm Hg) at 20°C
Grade ACS
Physical Form Liquid
Chemical Name or Material 2, 2, 4-Trimethylpentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.