missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10614641 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10614641 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10614641 Supplier JT Baker Supplier No. 8217.2500
  • ONLINE EXCLUSIVE PRICE
    223.00 €
    207.05 € / 2.5 L
    Save 15.95 €
    7% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Exceeds ACS Specifications

Specifications

Melting Point -107°C
CAS 540-84-1
Color Max. 10 (APHA)
Density 0.692 g/cm³ (20°C)
Quantity 2.5 L
Flash Point -12°C
Residue after Evaporation max. 0.001%
Assay Percent Range min. 99.0%
Molecular Formula C8H18
MDL Number MFCD00008943
Boiling Point 99.2°C (1013 hPa)
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.23
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.