missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, HPLC Grade, 99.7+%

Product Code. 11348127
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348127 1 L 1 L
11368127 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348127 Supplier Thermo Scientific Alfa Aesar Supplier No. 022901.K2
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.02% | CAS: 540-84-1 | C8H18 | 114.232 g/mol

2,2,4-Trimethylpentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point °C to 107°C
Color Colorless
Density 0.692
Boiling Point 98°C to 99°C
Flash Point -7°C (19°F)
Ignition Point 410°C (770°F)
Assay Percent Range >99.7%
Odor Nearly odorless
Refractive Index 1.391
Quantity 1 L
Beilstein 1696876
Solubility Information Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane and oils. Immiscible with water.
Formula Weight 114.23
Grade HPLC
Physical Form Liquid
Chemical Name or Material 2, 2, 4-Trimethylpentane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2,4-Trimethylpentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: 2,2,4-Trimethylpentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.