missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, for HPLC, Thermo Scientific

Product Code. 30280921 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280921 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280921 Supplier Thermo Scientific Supplier No. 268810050
  • ONLINE EXCLUSIVE PRICE
    252.00 €
    236.00 € / 5 L
    Save 16.00 €
    6% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 98°C
Quantity 5 L
Molecular Weight (g/mol) 114.23 g/mol
Percent Purity 99.50%
Grade HPLC
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.