missing translation for 'onlineSavingsMsg'
Learn More

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, 99%

Product Code. 11395169
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395169 1 g 1g
11305179 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395169 Supplier Thermo Scientific Alfa Aesar Supplier No. A18294.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19805-75-5 | C9H16N2O | 168.24 g/mol

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19805-75-5
Molecular Formula C9H16N2O
Molecular Weight (g/mol) 168.24
MDL Number MFCD00041846
InChI Key ACFYUJLIWIDSFM-UHFFFAOYSA-N
Synonym 2,2,5,5-tetramethyl-3-pyrroline-3-carboxamide, 2,2,5,5-tetramethyl-2,5-dihydro-1h-pyrrole-3-carboxamide, 1h-pyrrole-3-carboxamide, 2,5-dihydro-2,2,5,5-tetramethyl, 2,5-dihydro-2,2,5,5-tetramethyl-1h-pyrrole-3-carboxamide, 3-aminocarbonyl-2,2,5,5-tetramethy-3-pyrroline, maybridge3_000440, acmc-1c3ma, 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrroline, 3-aminocarbonyl-2,2,5,5-tetramethyl-3-pyrroline
PubChem CID 88255
IUPAC Name 2,2,5,5-tetramethyl-1H-pyrrole-3-carboxamide
SMILES CC1(C=C(C(N1)(C)C)C(=O)N)C

Specifications

Melting Point 179°C to 183°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 168.24
Percent Purity 99%
Chemical Name or Material 2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.