missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%

Product Code. 10573721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10573721 25 g Glass bottle 25g
11994821 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10573721 Supplier Thermo Scientific Acros Supplier No. 168320250
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1118-71-4 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-71-4
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
InChI Key YRAJNWYBUCUFBD-UHFFFAOYSA-N
Synonym dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497
PubChem CID 70700
IUPAC Name 2,2,6,6-tetramethylheptane-3,5-dione
SMILES CC(C)(C)C(=O)CC(=O)C(C)(C)C

Specifications

Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 72.0°C to 73.0°C (6.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CCOCH2COC(CH3)3
Refractive Index 1.4579 to 1.4599
Quantity 25 g
Beilstein 01,III,3149
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble.
Formula Weight 184.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
Show More Show Less
compliance-icons
Product Identifier
  • 2,2,6,6-Tetramethyl-3,5-heptanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.