missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-4-piperidone monohydrate, 99%

Product Code. 11463297
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463297 10 g 10g
11473297 50 g 50g
11483297 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463297 Supplier Thermo Scientific Alfa Aesar Supplier No. A16211.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10581-38-1 | C9H19NO2 | 173.256 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10581-38-1
Molecular Formula C9H19NO2
Molecular Weight (g/mol) 173.256
MDL Number MFCD00149393
InChI Key SFDODBGIDPNFEH-UHFFFAOYSA-N
Synonym 2,2,6,6-tetramethyl-4-piperidone hydrate, 2,2,6,6-tetramethyl-4-piperidone monohydrate, 4-piperidinone, 2,2,6,6-tetramethyl-, monohydrate, triacetone amine hydrate, acmc-20aokc, 2,2,6,6-tetramethyl-4-piperidonehydrate, 2,2,6,6-tetramethyl-4-oxopiperidine-hydrate, 2,2,6,6-tetramethylpiperidin-4-one, oxamethane, 2,2,6,6-tetramethylpiperidin-4-one-water 1/1, 4-piperidinone,2,2,6,6-tetramethyl-, hydrate 1:1
PubChem CID 22397602
IUPAC Name 2,2,6,6-tetramethylpiperidin-4-one;hydrate
SMILES CC1(CC(=O)CC(N1)(C)C)C.O

Specifications

Melting Point 63°C to 61°C
Quantity 10 g
Sensitivity Light sensitive
Formula Weight 173.26 (155.24 Anhydrous)
Percent Purity 99%
Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidone monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.