missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethyl)pyridine, 98%

Product Code. 11397515
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397515 1 g 1g
11307525 5 g 5g
11317525 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397515 Supplier Thermo Scientific Alfa Aesar Supplier No. L05627.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2706-56-1 | C7H10N2 | 122.17 g/mol

2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminoethyl)pyridine is used as active pharmaceutical ingredients.

Solubility
Soluble in water (partly), and most common polar solvent. Insoluble in non-polar solvents, and oils.

Notes
Air Sensitive. Store at 4°C. Stor in the dark. Incompatible with air, oxidizing agents, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2706-56-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006367
InChI Key XPQIPUZPSLAZDV-UHFFFAOYSA-N
Synonym 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine
PubChem CID 75919
ChEBI CHEBI:74024
SMILES NCCC1=CC=CC=N1

Specifications

Density 1.02
Boiling Point 208°C
Flash Point 100°C (212°F)
Odor Amine-like
Refractive Index 1.536
Quantity 1 g
UN Number UN2735
Beilstein 111208
Sensitivity Air Sensitive
Solubility Information Soluble in water (partly),and most common polar solvent. Insoluble in non-polar solvents,and oils.
IUPAC Name 2-pyridin-2-ylethanamine
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2-(2-Aminoethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.