missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Aminoethyl)thio]ethan-1-ol, ≥90%, Thermo Scientific™

Product Code. 10501893
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10501893 1 g 1g
10702967 5 g 5g
10744393 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10501893 Supplier Thermo Scientific Maybridge Supplier No. SB01634DA
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 24304-84-5
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.198
InChI Key ZRZOERYTFAWUQQ-UHFFFAOYSA-N
Synonym 2-2-aminoethyl thio ethanol, 2-2-aminoethyl sulfanyl ethanol, 2-2-aminoethyl thio ethan-1-ol, ethanol,2-2-aminoethyl thio, 2-2-aminoethylthio ethanol, 2-2-aminoethylthio ethan-1-ol, 5-amino-3-thiapentane-1-ol, 2-amino-2-hydroxydiethylsulfite, 2-2-aminoethylsulfanyl ethanol
PubChem CID 90457
IUPAC Name 2-(2-aminoethylsulfanyl)ethanol
SMILES C(CSCCO)N

Specifications

CAS Min % 90.0
Density 1.1200g/mL
Assay Percent Range 90%
Linear Formula NH2CH2CH2SCH2CH2OH
Quantity 1 g
Formula Weight 121.2
Percent Purity 90%
Chemical Name or Material 2-[(2-Aminoethyl)thio]ethan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.