missing translation for 'onlineSavingsMsg'
Learn More

2,2', Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis, MP Biomedicals™

Product Code. 11486791 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486791 1 g 1g
11496791 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486791 Supplier MP Biomedicals Supplier No. 0219502301
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 30931-67-0
Molecular Formula C18H24N6O6S4
Molecular Weight (g/mol) 548.67
MDL Number MFCD00010404,MFCD00010404
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
PubChem CID 91884754
IUPAC Name diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O

Specifications

Color Green
Quantity 1 g
Formula Weight 548.7
Physical Form Powder
Chemical Name or Material 2, 2' Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis
compliance-icons
Product Identifier
  • 2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic Acid) Diammonium Salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.