missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biimidazole

Product Code. 16647042 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16647042

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 492-98-8 | C6H6N4 | 134.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-98-8
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD00047014
InChI Key AZUHIVLOSAPWDM-UHFFFAOYSA-N
Synonym 2,2 '-Bisimidazole; 2,2 '-Diimidazole
IUPAC Name 1H,1'H-2,2'-biimidazole
SMILES N1C=CN=C1C1=NC=CN1

Specifications

Alpha Vector BIIMIDAZOLE
Melting Point >350°C
Color Gray to Yellow
Infrared Spectrum Conforms
Assay Percent Range 97.5 to 102.5 % (Titration with HClO4)
Packaging glass bottle
Quantity 1 g
UN Number 3265
Formula Weight 134.14
Physical Form Crystalline Powder
Chemical Name or Material 2,2 '-Biimidazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2'-Biimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2,2-Biimidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.