missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%

Product Code. 11405177
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405177 5 g 5g
11495167 1 g 1g
11415177 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405177 Supplier Thermo Scientific Alfa Aesar Supplier No. B22594.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6813-38-3 | C12H6N2O4 | 242.19 g/mol

2,2'-Bipyridine-4,4'-dicarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bipyridine-4,4′-dicarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6813-38-3
Molecular Formula C12H6N2O4
Molecular Weight (g/mol) 242.19
MDL Number MFCD00015430
InChI Key FXPLCAKVOYHAJA-UHFFFAOYSA-L
Synonym 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
PubChem CID 688094
IUPAC Name 2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
SMILES [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O

Specifications

Melting Point >310°C
Quantity 5 g
Solubility Information Insoluble in water. Soluble in common organic solvents.
Formula Weight 244.21
Percent Purity 98%
Chemical Name or Material 2,2'-Bipyridine-4,4'-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.