missing translation for 'onlineSavingsMsg'
Learn More

2,2′-Bipyridyl, ≥99.0% (NT), Honeywell Fluka™

Product Code. p-7120049 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
15661430 5 g Glass Bottle 5g
15651430 25 g Plastic Bottle 25g
15641430 100 g Glass Bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 15661430

Brand: Honeywell-Fluka 144545G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Redox indicator, for spectrophotometric det. of Fe,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2-pyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

Melting Point 69°C to 71°C
Boiling Point 273°C
UN Number UN2811
Beilstein 113089
Formula Weight 156.18g/mol
Percent Purity ≥99.0% (NT)
Packaging Glass Bottle
Quantity 5 g
Chemical Name or Material 2,2′-Bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.