missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%

Product Code. 11410707
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11410707 25 g 25g
11420707 100 g 100g
11430707 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11410707

Brand: Thermo Scientific Alfa Aesar B22515.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

Solubility
Soluble in water (209.2 g/L), at 20°C and alcohol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Melting Point 102°C to 105°C
Linear Formula (HOCH2)3CN(CH2CH2OH)2
Quantity 25 g
Beilstein 2205275
Solubility Information Soluble in water (209.2g/L),at 20°C and alcohol.
Formula Weight 209.24
Percent Purity ≥98%
Chemical Name or Material 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.