missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)propionic acid, 98+%

Product Code. 11358895
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358895 500 g 500g
11348895 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358895 Supplier Thermo Scientific Alfa Aesar Supplier No. L14014.36
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol

2,2-Bis(hydroxymethyl)propionic acid is used in the synthesis and characterization studies of polyurethane dispersion modified with polymethylsiloxane (Si-PUD) and its nanocomposites incorporated with clays1.2,2-Bis(hydroxymethyl)propionic acid based dendrimers have potent applications as delivery systems for potent anticancer drugs such as cisplatin and doxorubicin, as well as agents for both boron neutron capture therapy and photodynamic therapy Simple derivatives of this acid have been used to construct dendritic architectures via the click azide-acetylene cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Bis(hydroxymethyl)propionic acid is used in the synthesis and characterization studies of polyurethane dispersion modified with polymethylsiloxane (Si-PUD) and its nanocomposites incorporated with clays1.2,2-Bis(hydroxymethyl)propionic acid based dendrimers have potent applications as delivery systems for potent anticancer drugs such as cisplatin and doxorubicin, as well as agents for both boron neutron capture therapy and photodynamic therapy Simple derivatives of this acid have been used to construct dendritic architectures via the “click” azide-acetylene cycloaddition.

Solubility
Soluble in acetone (slightly), methanol, and water. Insoluble in benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4767-03-7
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00004199
InChI Key PTBDIHRZYDMNKB-UHFFFAOYSA-N
Synonym 2,2-bis hydroxymethyl propionic acid, dimethylolpropionic acid, 3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid, 2,2-bis hydroxymethyl propanoic acid, propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl, 2,2-dimethylolpropionic acid, dimethylol propionic acid, 2,2-bis hydroxymethyl propanoicacid, unii-4nhi8v17mn, 4nhi8v17mn
PubChem CID 78501
IUPAC Name 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
SMILES CC(CO)(CO)C(=O)O

Specifications

Melting Point 187°C to 191°C
Quantity 500 g
Beilstein 1756385
Sensitivity Hygroscopic
Merck Index 14,3253
Solubility Information Soluble in acetone (slightly),methanol,and water. Insoluble in benzene.
Formula Weight 134.13
Percent Purity ≥98%
Chemical Name or Material 2,2-Bis(hydroxymethyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.