missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)propionic acid, 98%

Product Code. 11955071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11955071 5 kg Plastic drum 5kg
10265050 500 g Plastic bottle 500g
10437730 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11955071 Supplier Thermo Scientific Acros Supplier No. 147970050
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4767-03-7
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
InChI Key PTBDIHRZYDMNKB-UHFFFAOYSA-N

Specifications

Melting Point 185.0°C to 190.0°C
Color White
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Linear Formula CH3C(CH2OH)2CO2H
Quantity 5 kg
Beilstein 03,401
Merck Index 15,3277
Synonym 6,7-dimethoxyquinoxaline-2,3 1h,4h-dione, 2,3-dihydroxy-6,7-dimethoxyquinoxaline, 6,7-dimethoxyquinoxaline-2,3-diol, chembl42430, 6,7-dimethoxy-1,4-dihydro-2,3-quinoxalinedione, acmc-20aogl, 2,3-quinoxalinedione,1,4-dihydro-6,7-dimethoxy, 6,7-dimethoxy-1,4-dihydro-quinoxaline-2,3-dione, 2,3-quinoxalinedione, 1,4-dihydro-6,7-dimethoxy, 6,7-dimethoxy-1,4-dihydro-2,3-quinoxalinedione monohydrate
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,slightly soluble in acetone,insoluble in benzene
SMILES CC(CO)(CO)C(O)=O
IUPAC Name 6,7-dimethoxy-1,4-dihydroquinoxaline-2,3-dione
PubChem CID 613780
Formula Weight 134.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2-Bis(hydroxymethyl)propionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,2-Bis(hydroxymethyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.