missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(hydroxymethyl)propionic Acid, TRC

Product Code. 30334183
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334183 100 g 100g
30338797 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30334183 Supplier Toronto Research Chemicals Supplier No. TRCB447448100G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Dimethylolpropionic acid
CAS 4767-03-7
Molecular Formula C5 H10 O4
Quantity 100 g
Synonym Propionic acid, 2,2-bis(hydroxymethyl)- (6CI,7CI,8CI), 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid, 2,2-Bis(hydroxymethyl)propanoic acid, 2,2-Bis(hydroxymethyl)propionic acid, 2,2-Bis(methylol)propionic acid, 2,2-Dihydroxymethylpropanoic acid, 2,2-Dimethylolpropionic acid, 3-Hydroxy-2-hydroxymethyl-2-methylpropionic acid, Bis(hydroxymethyl)propionic acid, Bis-MPA, DMPA, Dimethylolpropionic acid, NSC 96616, Nikkamer PA, Propanoic acid, 2,2-bis(hydroxymethyl)-, α,α-Bis(hydroxymethyl)propionic acid, α,α-Dimethylolpropionic acid, 2,2-Dimethylolpropionic Acid
InChI Formula InChI=1S/C5H10O4/c1-5(2-6,3-7)4(8)9/h6-7H,2-3H2,1H3,(H,8,9)
SMILES CC(CO)(CO)C(=O)O
IUPAC Name 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
Molecular Weight (g/mol) 134.1305
Formula Weight 134.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.