missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bithiophene-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10531682
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10531682 250 mg 250mg
10212542 1 g 1g
10674745 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10531682 Supplier Thermo Scientific Maybridge Supplier No. CC14601CB
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2060-55-1
Molecular Formula C9H6O2S2
Molecular Weight (g/mol) 210.265
InChI Key PTNWHEHDBNNKEO-UHFFFAOYSA-N
Synonym 2,2'-bithiophene-5-carboxylic acid, 5-thiophen-2-yl thiophene-2-carboxylic acid, 2,2-bithiophene-5-carboxylic acid, 2,2'-bithiophene-5-carboxylicacid, 2,2' bithiophenyl-5-carboxylic acid, 2,2'-bithiophene-5-ca, 5-carboxy-2,2'-bithiophene, 2,2'-bithienyl-5'-carboxylic, 5-2-thienyl-2-thiophenecarboxylic acid
PubChem CID 150965
IUPAC Name 5-thiophen-2-ylthiophene-2-carboxylic acid
SMILES C1=CSC(=C1)C2=CC=C(S2)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 210.28
Percent Purity 97%
Chemical Name or Material 2, 2'-Bithiophene-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.