missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethyl)-1,3-dioxolane, 96%

Product Code. 10236890 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236890 10 g Glass bottle 10g
10096133 25 g Glass bottle 25g
10195170 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10236890 Supplier Thermo Scientific Acros Supplier No. 213540100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18742-02-4 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18742-02-4
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00003216
InChI Key GGZQLTVZPOGLCC-UHFFFAOYSA-N
Synonym 2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan
PubChem CID 87776
IUPAC Name 2-(2-bromoethyl)-1,3-dioxolane
SMILES BrCCC1OCCO1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.5400g/mL
Boiling Point 68°C to 70°C (8.0 mmHg)
Flash Point 65°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4783 to 1.4803
Quantity 10 g
Fieser 07,37; 11,78
Specific Gravity 1.54
Solubility Information Solubility in water: immiscible
Formula Weight 181.03
Percent Purity 96%
Chemical Name or Material 2-(2-Bromoethyl)-1, 3-dioxolane
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Bromoethyl)-1,3-dioxolane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.