missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Butoxyethoxy)ethanol, 99+%

Product Code. 10763144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763144 2.5 L Glass bottle 2.5 L
10775041 1 L Glass bottle 1 L
11984721 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10763144 Supplier Thermo Scientific Acros Supplier No. 149710025
  • ONLINE EXCLUSIVE PRICE
    81.90 €
    79.50 € / 2.5 L
    Save 2.40 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-34-5 | C8H18O3 | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-34-5
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.23
InChI Key OAYXUHPQHDHDDZ-UHFFFAOYSA-N
Synonym 2-2-butoxyethoxy ethanol, diethylene glycol monobutyl ether, butyldiglycol, butyl carbitol, diethylene glycol butyl ether, butyl diglycol, butyl digol, ethanol, 2-2-butoxyethoxy, butoxyethoxyethanol, butoxydiglycol
PubChem CID 8177
IUPAC Name 2-(2-butoxyethoxy)ethanol
SMILES CCCCOCCOCCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -68.0°C
Density 0.9550g/mL
Boiling Point 231.0°C
Flash Point 100°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3OCH2CH2OCH2CH2OH
Refractive Index 1.4310 to 1.4330
Quantity 2.5 L
Beilstein 01,IV,2394
Merck Index 15,3133
Specific Gravity 0.955
Solubility Information Solubility in water: soluble. Other solubilities: miscible with oils, miscible with most common organic solvents
Viscosity 6.16 mPa.s (20°C)
Formula Weight 162.23
Percent Purity 99+%
Chemical Name or Material 2-(2-Butoxyethoxy)ethanol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Butoxyethoxy)ethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.