missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Butoxyethoxy)ethyl acetate, ≥99.2%, Honeywell™

Product Code. 15625780 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15625780 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15625780 Supplier Honeywell Research Chemicals Supplier No. 5375351L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

≥99.2%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 124-17-4
Melting Point -31.99°C
Color Yellow
Density 0.978g/mL at 25°C
Boiling Point 245°C
Explosion Limit 10.7%, (203°F), 0.76% (135°F)
Flash Point 102°C (215.6°F)
Ignition Point 393°F
Packaging Glass Bottle
Molecular Formula C10H20O4
Linear Formula CH3COOCH2CH2OCH2CH2O(CH2)3CH3
Refractive Index n20/D 1.423
MDL Number MFCD00009458
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 1771533
Synonym 2-2-butoxyethoxy ethyl acetate, butoxyethoxyethyl acetate, diethylene glycol monobutyl ether acetate, butyl diglycol acetate, butyl carbitol acetate, ektasolve db acetate, 2-2-butoxyethoxy ethanol acetate, butyl diethylene glycol acetate, butylkarbitolacetat, diglycol monobutyl ether acetate
InChI Key VXQBJTKSVGFQOL-UHFFFAOYSA-N
SMILES CCCCOCCOCCOC(=O)C
IUPAC Name 2-(2-butoxyethoxy)ethyl acetate
Molecular Weight (g/mol) 204.266
PubChem CID 31288
Formula Weight 204.26g/mol
Percent Purity ≥99.2%
Chemical Name or Material 2-(2-Butoxyethoxy)ethyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.