missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chloroethoxy)ethanol, TRC

Product Code. 30008669
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008669 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008669 Supplier Toronto Research Chemicals Supplier No. TRCC36574050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-89-7
Molecular Formula C4 H9 Cl O2
Molecular Weight (g/mol) 124.566
Synonym Ethanol, 2-(2-chloroethoxy)-, 2-(2-Chloroethoxy)ethanol, 2-(2-Hydroxyethoxy)ethyl chloride, 2-(2'-Chloroethoxy)ethanol, 2-(Chloroethoxy)ethanol, 2-Chloroethyl 2-hydroxyethyl ether, 2-[(2-Chloroethyl)oxy]ethanol, 5-Chloro-3-oxa-1-pentanol, Diethylene glycol monochlorohydrin, Diglycol chlorohydrin, Ethylene glycol mono(2-chloroethyl) ether, NSC 2648
IUPAC Name 2-(2-chloroethoxy)ethanol
SMILES OCCOCCCl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C4H9ClO2/c5-1-3-7-4-2-6/h6H,1-4H2
Formula Weight 124.0291
Chemical Name or Material 2-(2-Chloroethoxy)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.