missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-chloroethyl)sulfonyl]-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 10421553
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10421553 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10421553 Supplier Thermo Scientific Maybridge Supplier No. BTB07960DA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 243643-97-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H7ClF3NO2S
MDL Number MFCD00099609
Quantity 1 g
Synonym 2-2-chloroethyl sulfonyl-5-trifluoromethyl pyridine, 2-2-chloroethanesulfonyl-5-trifluoromethyl pyridine, 2-2-chloroethylsulfonyl-5-trifluoromethyl pyridine, 2-2-chloroethyl sulphonyl-5-trifluoromethyl pyridine, pyridine,2-2-chloroethyl sulfonyl-5-trifluoromethyl
InChI Key VACOIIVHOIMMEF-UHFFFAOYSA-N
SMILES C1=CC(=NC=C1C(F)(F)F)S(=O)(=O)CCCl
IUPAC Name 2-(2-chloroethylsulfonyl)-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol) 273.654
PubChem CID 2779640
Formula Weight 273.66
Percent Purity 97%
Chemical Name or Material 2-[(2-Chloroethyl)sulfonyl]-5-(trifluoromethyl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.