missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2,2'-Dibromo-1,1'-binaphthyl, 96%

Product Code. 11441620
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441620 1 g 1g
11451620 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441620 Supplier Thermo Scientific Alfa Aesar Supplier No. H26987.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 74866-28-7 | C20H12Br2 | 412.124 g/mol

The synthesis of this class of compounds relies on the lithiation of racemic 2,2'-dibromo-1,1'-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The synthesis of this class of compounds relies on the lithiation of racemic 2,2′-dibromo-1,1′-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.

Solubility
Sparingly Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74866-28-7
Molecular Formula C20H12Br2
Molecular Weight (g/mol) 412.124
MDL Number MFCD00188005
InChI Key IJUDEFZBMMRSNM-UHFFFAOYSA-N
Synonym 2,2'-dibromo-1,1'-binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, racemic-2,2'-dibromo-1,1'-binaphthyl, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, s---2,2-dibromo-1,1'-binaphthyl, 2,2'-dibromo 1,1' binaphthyl, 1,1'-bi 2-bromonaphthalene
PubChem CID 619909
IUPAC Name 2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br

Specifications

Boiling Point 178°C to 183°C
Quantity 1 g
Solubility Information Sparingly Soluble in water.
Formula Weight 412.23
Percent Purity 96%
Chemical Name or Material (+/-)-2,2'-Dibromo-1,1'-binaphthyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.