missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dibromo-4'-fluoroacetophenone, 98%

Product Code. 11379745
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379745 1 g 1g
11389745 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379745 Supplier Thermo Scientific Alfa Aesar Supplier No. L19788.03
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7542-64-5 | C8H5Br2FO | 295.933 g/mol

2,2-Dibromo-4'-fluoroacetophenone is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dibromo-4′-fluoroacetophenone is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7542-64-5
Molecular Formula C8H5Br2FO
Molecular Weight (g/mol) 295.933
MDL Number MFCD03791309
InChI Key XFFGHBAVJPOWDG-UHFFFAOYSA-N
Synonym 2,2-dibromo-1-4-fluorophenyl ethanone, alpha,alpha-dibromo-4-fluoroacetophenone, 2,2-dibromo-4'-fluoroacetophenone, 4'-fluoro-alpha,alpha-dibromoacetophenone, 2,2-dibromo-1-4-fluorophenyl ethan-1-one, 2,2-bis bromanyl-1-4-fluorophenyl ethanone
PubChem CID 2783255
IUPAC Name 2,2-dibromo-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)C(Br)Br)F

Specifications

Melting Point 36°C to 38°C
Quantity 1 g
UN Number UN3261
Beilstein 2256317
Solubility Information Insoluble in water.
Formula Weight 295.93
Percent Purity 98%
Chemical Name or Material 2,2-Dibromo-4'-fluoroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.