missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dichloroacetic Acid-1-13C, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
Specifications
Specifications
| Chemical Name or Material | 2,2-Dichloroacetic Acid-1-13C |
| CAS | 173470-70-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13CH2Cl2O2 |
| Quantity | 2.5 mg |
| Synonym | Dichloroacetic acid 13C (C1-13C), Acetic-1-13C acid, 2,2-dichloro- (9CI) |
| InChI Formula | InChI=1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i2+1 |
| SMILES | O[13C](C(Cl)Cl)=O |
| Molecular Weight (g/mol) | 129.93 |
| Percent Purity | >95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction