missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diethoxyethanol, TRC

Product Code. 30369422
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369422 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369422 Supplier Toronto Research Chemicals Supplier No. TRCD4419401G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 621-63-6
Molecular Formula C6H14O3
Quantity 1 g
Synonym Diethyl Acetal Glycolaldehyde, 2,2-Diethoxyethanol, 2-Hydroxyacetaldehyde Diethyl Acetal, Hydroxyacetaldehyde Diethyl Acetal, NSC 9255
InChI Formula InChI=1S/C6H14O3/c1-3-8-6(5-7)9-4-2/h6-7H,3-5H2,1-2H3
SMILES CCOC(CO)OCC
IUPAC Name 2,2-diethoxyethanol
Molecular Weight (g/mol) 134.17
Formula Weight 134.0943
Chemical Name or Material 2,2-Diethoxyethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.