missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde, 97%

Product Code. 11491267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491267 1 g 1g
11401277 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491267 Supplier Thermo Scientific Alfa Aesar Supplier No. B24232.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 656-42-8 | C8H4F2O3 | 186.114 g/mol

2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde is used as primary and secondary intermediate.

Solubility
Sparingly soluble in water (0.31 g/L at 25°C).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 656-42-8
Molecular Formula C8H4F2O3
Molecular Weight (g/mol) 186.114
MDL Number MFCD00792420
InChI Key GGERGLKEDUUSAP-UHFFFAOYSA-N
Synonym 2,2-difluorobenzo d 1,3 dioxole-5-carbaldehyde, 2,2-difluorobenzodioxole-5-carboxaldehyde, 2,2-difluoro-1,3-benzodioxole-5-carboxaldehyde, 2,2-difluoro-2h-1,3-benzodioxole-5-carbaldehyde, 2,2-difluoro-5-formylbenzodioxole, 2,2-difluoro-5-formyl-1,3-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole-5-carbaldehyde, 5-formyl-2,2-difluorobenzodioxole, 1,3-benzodioxole-5-carboxaldehyde, 2,2-difluoro, 2,2-difluoro-1,3-benzodioxole-5-arboxaldehyde
PubChem CID 2736973
IUPAC Name 2,2-difluoro-1,3-benzodioxole-5-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)OC(O2)(F)F

Specifications

Density 1.399
Boiling Point 103°C to 105°C (20 mmHg)
Refractive Index 1.499
Quantity 1 g
Beilstein 1369253
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water (0.31g/L at 25°C).
Formula Weight 186.11
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.