missing translation for 'onlineSavingsMsg'
Learn More

2,2-difluoro-3-phenylpropanoic acid, TRC

Product Code. 30380893
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380893 25 mg -
30369988 2.5 mg 2.50mg
30386364 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380893 Supplier Toronto Research Chemicals Supplier No. TRCD45517325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142977-64-8
Molecular Formula C9H8O2F2
Molecular Weight (g/mol) 186.16
SMILES OC(=O)C(F)(F)Cc1ccccc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H8F2O2/c10-9(11,8(12)13)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,12,13)
Formula Weight 186.0492
Chemical Name or Material 2,2-difluoro-3-phenylpropanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.