missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-4-pentenoic acid, 97%

Product Code. p-7031192
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15444867

Brand: Thermo Scientific Alfa Aesar H32173.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 55039-89-9 | C5H6F2O2 | 136.098 g/mol

2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55039-89-9
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD09800642
InChI Key LHOKYUDUAYXFGF-UHFFFAOYSA-N
Synonym 2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n
PubChem CID 11954656
IUPAC Name 2,2-difluoropent-4-enoic acid
SMILES C=CCC(C(=O)O)(F)F

Specifications

Boiling Point 56°C to 58°C (3 mmHg)
Quantity 250 mg
UN Number UN3265
Solubility Information Immiscible with water.
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-4-pentenoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.