missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-N-methoxy-N-methylacetamide, 95%

Product Code. 11435560
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435560 5 g 5g
15403317 250 mg 250mg
11425560 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435560 Supplier Thermo Scientific Alfa Aesar Supplier No. H25833.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 142492-01-1 | C4H7F2NO2 | 139.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142492-01-1
Molecular Formula C4H7F2NO2
Molecular Weight (g/mol) 139.10
MDL Number MFCD06248152
InChI Key CUPRFYMJGQMIIC-UHFFFAOYSA-N
PubChem CID 19023731
IUPAC Name 2,2-difluoro-N-methoxy-N-methylacetamide
SMILES CON(C)C(=O)C(F)F

Specifications

Boiling Point 40°C to 41°C (0.7 mmHg)
Refractive Index 1.402
Quantity 5 g
Synonym n-methyl-n-methoxydifluoroacetamide, acetamide, 2,2-difluoro-n-methoxy-n-methyl, acmc-20a3nv, n-methoxy-n-methyldifluoroacetamide, n-methoxy-n-methyl difluoroacetamide, n-difluoroacetyl-n-methoxymethylamine, acetamide,2,2-difluoro-n-methoxy-n-methyl, 2,2-bis fluoranyl-n-methoxy-n-methyl-ethanamide
Formula Weight 139.1
Percent Purity 95%
Chemical Name or Material 2,2-Difluoro-N-methoxy-N-methylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.