missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorobutyric acid, 97%

Product Code. 15483307
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15483307

Brand: Thermo Scientific Alfa Aesar H25830.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2366-62-3 | C4H6F2O2 | 124.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2366-62-3
Molecular Formula C4H6F2O2
Molecular Weight (g/mol) 124.087
MDL Number MFCD03093765
InChI Key SBSKRVZFWQPJID-UHFFFAOYSA-N
Synonym 2,2-difluorobutyric acid, 2,2-difluorobutyricacid, pubchem15470, difluoramino butyric acid, 2,2-bis fluoranyl butanoic acid
PubChem CID 3711426
IUPAC Name 2,2-difluorobutanoic acid
SMILES CCC(C(=O)O)(F)F

Specifications

Melting Point 7°C to 9°C
Density 1.22
Boiling Point 100°C to 101°C (8.5 mmHg)
Linear Formula CH3CH2CF2COOH
Refractive Index 1.3645
Quantity 250 mg
UN Number UN3265
Formula Weight 124.09
Percent Purity 97%
Chemical Name or Material 2,2-Difluorobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.