missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoropropionic acid, 97%, Thermo Scientific Chemicals

Product Code. 11331959
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331959 1 g 1g
15433347 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331959 Supplier Thermo Scientific Alfa Aesar Supplier No. H26030.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-96-6
Molecular Formula C3H4F2O2
Molecular Weight (g/mol) 110.06
MDL Number MFCD03093764
InChI Key PMWGIVRHUIAIII-UHFFFAOYSA-N
Synonym 2,2-difluoropropionic acid, 2,2-difluoropropionicacid, 2,2-difluoro-propionic acid, propanoic acid, 2,2-difluoro, acmc-1ctsu, 2,2-difluorpropionic acid
PubChem CID 2782825
IUPAC Name 2,2-difluoropropanoic acid
SMILES CC(F)(F)C(O)=O

Specifications

Melting Point 36°C to 44°C
Boiling Point 142°C
Flash Point 66°C (150°F)
Linear Formula CH3CF2COOH
Quantity 1 g
UN Number UN3261
Formula Weight 110.06
Percent Purity 97%
Chemical Name or Material 2,2-Difluoropropionic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.