missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorosuccinic acid, 94%

Product Code. 11330204
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11330204

Brand: Thermo Scientific Alfa Aesar L00773.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 665-31-6 | C4H4F2O4 | 154.07 g/mol

2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 665-31-6
Molecular Formula C4H4F2O4
Molecular Weight (g/mol) 154.07
MDL Number MFCD00040970
InChI Key ZYLFHISLYSHWRH-UHFFFAOYSA-N
Synonym 2,2-difluorosuccinic acid, 2,2-difluorosuccinicacid, butanedioic acid, 2,2-difluoro, pubchem23428, 2,2-difluoro-butanedioic acid, butanedioic acid,2,2-difluoro, 2,2-difluoro succinic acid, 2,2-difluorobutane-1,4-dioic acid
PubChem CID 69581
IUPAC Name 2,2-difluorobutanedioic acid
SMILES OC(=O)CC(F)(F)C(O)=O

Specifications

Melting Point 139°C to 144°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Beilstein 1773355
Solubility Information Soluble in water.
Formula Weight 154.07
Percent Purity 94%
Chemical Name or Material 2,2-Difluorosuccinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.