missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Difluorosuccinic acid, 94%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, and strong bases.
2,2-Difluorosuccinic acid is used in the preparation of fluorofumaric acid. It is also used in enolate alkylation.
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, and strong bases.
Specifications
Specifications
| Melting Point | 139°C to 144°C |
| Flash Point | >110°C (230°F) |
| Quantity | 1 g |
| UN Number | UN3261 |
| Beilstein | 1773355 |
| Solubility Information | Soluble in water. |
| Formula Weight | 154.07 |
| Percent Purity | 94% |
| Chemical Name or Material | 2,2-Difluorosuccinic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction