missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxypropane, 98+%

Product Code. 10274330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
1 L
10 L
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Packaging Conditionnement
10274330 500 mL Glass bottle 500mL
10214290 1 L Glass bottle 1 L
10410384 2.5 L Glass bottle 2.5 L
11994761 10 L Metal Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 10274330 Fournisseur Thermo Scientific Acros Code fournisseur 115635000
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 77-76-9 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 77-76-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
InChI Key HEWZVZIVELJPQZ-UHFFFAOYSA-N
Synonym acetone dimethyl acetal, propane, 2,2-dimethoxy, acetone dimethyl ketal, acetone, dimethyl acetal, acetone-dimethyl acetal, 2,2-dimethoxy propane, acetone dimethylacetal, 2,2-dimethyoxypropane, 2,2-dimethoxy-propane, dimethoxypropan
PubChem CID 6495
IUPAC Name 2,2-dimethoxypropane
SMILES CC(C)(OC)OC

Spécification

CAS Min % 98.0
CAS Max % 100.0
Melting Point -47.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 83.0°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C(OCH3)2
Refractive Index 1.3760 to 1.3780
Quantity 500 mL
Beilstein 01,648
Fieser 01,268; 05,226; 13,199; 15,138
Specific Gravity 0.84
Solubility Information Solubility in water: 18 g/100g (25°C). Other solubilities: soluble in acetone, benzene, ccl4, ether,, n-heptane and methanol
Viscosity 0.452 mPa.s (25°C)
Formula Weight 104.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2,2-Dimethoxypropane
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • 2,2-Dimethoxypropane
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P310-Immediately call a POISON CENTER/doctor/.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.