missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxan-5-one, tech. 90%, Thermo Scientific Chemicals

Product Code. 11486890
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486890 1 g 1g
15479129 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486890 Supplier Thermo Scientific Alfa Aesar Supplier No. L20177.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74181-34-3 | Molecular Formula: C6H10O3 | Molecular Weight: 130.143 g/mol

2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with oxidizing agents. Air, moisture and heat sensitive.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74181-34-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00671514
InChI Key ASFQDNDZFGFMMP-UHFFFAOYSA-N
Synonym 2,2-dimethyl-1,3-dioxane-5-one, 2,2-dimethyl-1,3 dioxan-5-one, 1,3-dioxan-5-one, 2,2-dimethyl, acmc-1bg7h, ksc496q3n, 5-oxo-2,2-dimethyl-1,3-dioxane, 2,2-dimethyl-1,3-dioxan-5-one, technical 250mg
PubChem CID 2733141
IUPAC Name 2,2-dimethyl-1,3-dioxan-5-one
SMILES CC1(OCC(=O)CO1)C

Specifications

Boiling Point 40°C to 46°C (14 mmHg)
Refractive Index 1.4315
Quantity 1 g
Beilstein 3061933
Sensitivity Air and moisture sensitive
Solubility Information Miscible with water and most organic solvents.
Formula Weight 130.15
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,2-Dimethyl-1,3-dioxan-5-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.