missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxolane, 98+%

Product Code. 11380064
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380064 25 g 25g
11370064 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380064 Supplier Thermo Scientific Alfa Aesar Supplier No. L00482.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2916-31-6 | C5H10O2 | 102.13 g/mol

2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Insoluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2916-31-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00014106
InChI Key SIJBDWPVNAYVGY-UHFFFAOYSA-N
PubChem CID 76209
IUPAC Name 2,2-dimethyl-1,3-dioxolane
SMILES CC1(C)OCCO1

Specifications

Density 0.946
Boiling Point 91°C to 93°C
Flash Point 6°C (42°F)
Refractive Index 1.398
Quantity 25 g
UN Number UN1993
Beilstein 102775
Synonym 1,3-dioxolane, 2,2-dimethyl, 2,2-dimethyl-1,3-dioxolan, 1,3-dioxolane, dimethyl, acetone ethylene ketal, acetone ethylene acetal, acetone, cyclic ethylene acetal, 1,3-dioxolane,2,2-dimethyl, sijbdwpvnayvgy-uhfffaoysa, 2,2-dimethyl-1,3-dioxolane
Solubility Information Insoluble in water.
Formula Weight 102.13
Percent Purity ≥98%
Chemical Name or Material 2,2-Dimethyl-1,3-dioxolane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.