missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-propanediol, 99%

Product Code. 11474123
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474123 2500 g 2500g
11494120 250 g 250g
11464123 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474123 Supplier Thermo Scientific Alfa Aesar Supplier No. A10340.0E
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 126-30-7 | C5H12O2 | 104.15 g/mol

Used for protection of carbonyl groups as 4,4-dimethyl-1,3-dioxanes, often preferred to the more usual 1,3-dioxolanes due to easier separation and greater stability, for protection of carbonyl group of 4-bromo-2-propanone using p-toluenesulfonic acid in the presence of triethyl orthoformate. It is used in the synthesis of polyesters, paints, lubricants, and plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for protection of carbonyl groups as 4,4-dimethyl-1,3-dioxanes, often preferred to the more usual 1,3-dioxolanes due to easier separation and greater stability, for protection of carbonyl group of 4-bromo-2-propanone using p-toluenesulfonic acid in the presence of triethyl orthoformate. It is used in the synthesis of polyesters, paints, lubricants, and plasticizers.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents, water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-30-7
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00004685
InChI Key SLCVBVWXLSEKPL-UHFFFAOYSA-N
Synonym neopentyl glycol, 2,2-dimethyl-1,3-propanediol, dimethylolpropane, neopentylene glycol, neopentanediol, neopentylglycol, 1,3-propanediol, 2,2-dimethyl, neol, hydroxypivalyl alcohol, dimethyltrimethylene glycol
PubChem CID 31344
IUPAC Name 2,2-dimethylpropane-1,3-diol
SMILES CC(C)(CO)CO

Specifications

Melting Point 125°C to 130°C
Density 1.042
Boiling Point 210°C to 212°C
Flash Point 103°C (217°F)
Odor Mild
Quantity 2500 g
Beilstein 605291
Sensitivity Hygroscopic
Merck Index 14,6458
Solubility Information Soluble in water.
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material 2,2-Dimethyl-1,3-propanediol
Show More Show Less
compliance-icons
Product Identifier
  • 2,2-Dimethyl-1,3-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,2-Dimethyl-1,3-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.