missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-propanediol, 99%

Product Code. 10301451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10301451 500 g Glass bottle 500g
10741442 25 g Glass bottle 25g
10163663 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10301451 Supplier Thermo Scientific Acros Supplier No. 408555000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 126-30-7 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-30-7
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00004685
InChI Key SLCVBVWXLSEKPL-UHFFFAOYSA-N
Synonym neopentyl glycol, 2,2-dimethyl-1,3-propanediol, dimethylolpropane, neopentylene glycol, neopentanediol, neopentylglycol, 1,3-propanediol, 2,2-dimethyl, neol, hydroxypivalyl alcohol, dimethyltrimethylene glycol
PubChem CID 31344
IUPAC Name 2,2-dimethylpropane-1,3-diol
SMILES CC(C)(CO)CO

Specifications

Acidity 0.1mg KOH/g max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point 122.0°C to 128.0°C
Color White
Boiling Point 208.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2C(CH3)2CH2OH
Quantity 500 g
Beilstein 01, IV, 2551
Fieser 05,259
Merck Index 15, 6543
Solubility Information Solubility in water: 830g/L (20°C). Other solubilities: freely soluble in alcohol and ether
Formula Weight 104.15
Percent Purity 99%
Physical Form Crystalline Chunks
Chemical Name or Material 2, 2-Dimethyl-1, 3-propanediol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,2-Dimethyl-1,3-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,2-Dimethyl-1,3-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.